Få mere at vide
Alpha-Spinasterol, TRC
Artikelnummer. 30136033
Skift visning
Mængde:
1 mg
5 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30136033 25 mg
30006377 1 mg
30012787 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30136033 Leverandør Toronto Research Chemicals Leverandørnr. TRCS68190025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 481-18-5
Molekylær formel C29 H48 O
Molekylvægt (g/mol) 412.69
Synonym Stigmasta-7,22-dien-3-ol, (3β,5α,22E)-, 5α-Stigmasta-7,22-dien-3β-ol (6CI,7CI), 5α-Stigmasta-7,22-dien-3β-ol, (E)- (8CI), (3β,5α,22E)-Stigmasta-7,22-dien-3-ol, Bessisterol, Hitodesterol, Spinasterol, Stigmasta-7,22-dien-3β-ol, δ7,22-Stigmastan-3β-ol, α-Spinasterin, α-Spinasterol
IUPAC navn (3S,5S,9R,10S,13R,14R,17R)-17-[(E,2R,5S)-5-ethyl-6-methylhept-3-en-2-yl]-10,13-dimethyl-2,3,4,5,6,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-ol
SMIL CC[C@H](\C=C\[C@@H](C)[C@H]1CC[C@H]2C3=CC[C@H]4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C)C(C)C
Tekniske data
Kemisk navn eller materiale A-Spinasterol
Mængde 25 mg
InChI formel InChI=1S/C29H48O/c1-7-21(19(2)3)9-8-20(4)25-12-13-26-24-11-10-22-18-23(30)14-16-28(22,5)27(24)15-17-29(25,26)6/h8-9,11,19-23,25-27,30H,7,10,12-18H2,1-6H3/b9-8+/t20-,21-,22+,23+,25-,26+,27+,28+,29-/m1/s1
Formel vægt 412.3705