missing translation for 'onlineSavingsMsg'
Få mere at vide

(-)-Alpha-Santalene, TRC

Artikelnummer. 30134959
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 mg
2.5 mg
5 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30134959 1 mg
17998584 2.5 mg
17999634 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30134959 Leverandør Toronto Research Chemicals Leverandørnr. TRCS1150651MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 512-61-8
Molekylær formel C15H24
Molekylvægt (g/mol) 204.35
Synonym (2R,6S,7S)-1,7-Dimethyl-7-(4-methyl-3-penten-1-yl)tricyclo[2.2.1.02,6]heptane, Santalene
IUPAC navn (2R,6S,7S)-1,7-dimethyl-7-(4-methylpent-3-enyl)tricyclo[2.2.1.02,6]heptane
SMIL CC(=CCC[C@@]1(C)C2C[C@@H]3C(C2)C13C)C

Specifications

Kemisk navn eller materiale (-)-alpha-Santalene
Renhedsgrad noter HPLC
Mængde 1 mg
InChI formel InChI=1S/C15H24/c1-10(2)6-5-7-14(3)11-8-12-13(9-11)15(12,14)4/h6,11-13H,5,7-9H2,1-4H3/t11?,12-,13+,14-,15-/m0/s1
Formel vægt 204.1878
Procent renhed >95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.