Learn More
alpha-Pyrone, 97%, BLD Pharm™
Specifications
Specifications
| CAS | 504-31-4 |
| Molecular Formula | C5H4O2 |
| MDL Number | MFCD00006639 |
| Quantity | 1 g |
| SMILES | O=C1C=CC=CO1 |
| Molecular Weight (g/mol) | 96.08 |
| Percent Purity | 97% |
| Chemical Name or Material | alpha-Pyrone |