Få mere at vide
Alpha-Phenyl-Alpha-(2-pyridyl)acetonitrile-d4, TRC
Artikelnummer. 30411246
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30411246 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30411246 Leverandør Toronto Research Chemicals Leverandørnr. TRCP3366525MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale alpha-Phenyl-alpha-(2-pyridyl)acetonitrile-d4
CAS 1217002-29-3
Molekylær formel C13H6D4N2
Mængde 5 mg
Synonym alpha-Phenyl-2-pyridineacetonitrile-d4, 2-Pyridylphenylacetonitrile-d4, NSC 16276-d4, Phenyl(pyridin-2-yl)acetonitrile-d4
InChI formel InChI=1S/C13H10N2/c14-10-12(11-6-2-1-3-7-11)13-8-4-5-9-15-13/h1-9,12H/i4D,5D,8D,9D
SMIL [2H]c1nc(C(C#N)c2ccccc2)c([2H])c([2H])c1[2H]
IUPAC navn 2-phenyl-2-(3,4,5,6-tetradeuteriopyridin-2-yl)acetonitrile
Molekylvægt (g/mol) 198.26
Formel vægt 198.1095