Få mere at vide
Alpha-Phenyl-Alpha-(2-propyn-1-yloxy)benzeneacetic Acid, TRC
Artikelnummer. 30423686
Skift visning
Mængde:
100 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30423686 250 mg
30413266 1 g
30413581 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30423686 Leverandør Toronto Research Chemicals Leverandørnr. TRCP336405250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 2766-01-0
Molekylær formel C17H14O3
Molekylvægt (g/mol) 266.29
Synonym Diphenyl(2-propynyloxy)acetic Acid, Propinoxiphenyl Mandelic Acid
IUPAC navn 2,2-diphenyl-2-prop-2-ynoxyacetic acid
SMIL OC(=O)C(OCC#C)(c1ccccc1)c2ccccc2
Tekniske data
Kemisk navn eller materiale alpha-Phenyl-alpha-(2-propyn-1-yloxy)benzeneacetic Acid
Mængde 250 mg
InChI formel InChI=1S/C17H14O3/c1-2-13-20-17(16(18)19,14-9-5-3-6-10-14)15-11-7-4-8-12-15/h1,3-12H,13H2,(H,18,19)
Formel vægt 266.0943