Learn More
Alpha,O-Dimethyl Serotonin Hydrochloride, TRC
Product Code. 30388360
Change view
Click to view available options
Quantity:
250 mg
2.5 g
Unit Size:
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30388360 2.5 g 2.5g
30385819 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30388360 Supplier Toronto Research Chemicals Supplier No. TRCD4792252.5G
 Restricted Item
Forventet forsendelse: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1016-44-0
Molecular Formula C12 H16 N2 O . Cl H
Molecular Weight (g/mol) 240.729
Synonym 5-Methoxy-α-methyl-1H-indole-3-ethanamine Hydrochloride (1:1), 5-Methoxy-α-methyltryptamine Hydrochloride, 3-(2-Aminopropyl)-5-methoxyindole Hydrochloride (6CI,7CI,8CI)
IUPAC Name 1-(5-methoxy-1H-indol-3-yl)propan-2-amine;hydrochloride
SMILES Cl.COc1ccc2[nH]cc(CC(C)N)c2c1
Specifications
Quantity 2.5 g
InChI Formula InChI=1S/C12H16N2O.ClH/c1-8(13)5-9-7-14-12-4-3-10(15-2)6-11(9)12;/h3-4,6-8,14H,5,13H2,1-2H3;1H
Formula Weight 240.103
Chemical Name or Material 5-Methoxy-alpha-methyltryptamine Hydrochloride