missing translation for 'onlineSavingsMsg'
Få mere at vide

Alfa, O-Dimethyl Serotonin Hydrochlorid, TRC

Artikelnummer. 30385819
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
250 mg
2.5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30385819 250 mg
30388360 2.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30385819 Leverandør Toronto Research Chemicals Leverandørnr. TRCD479225250MG
 Begrænset vare
Forventet forsendelse: 10-12-2026
Læg i indkøbskurv
Læg i indkøbskurv

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy

Højrenheds organiske molekyler og analytiske standarder, strategisk leveret verden over for at muliggøre innovation og kommerciel succes.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1016-44-0
Molekylær formel C12 H16 N2 O . Cl H
Molekylvægt (g/mol) 240.729
Synonym 5-Methoxy-α-methyl-1H-indole-3-ethanamine Hydrochloride (1:1), 5-Methoxy-α-methyltryptamine Hydrochloride, 3-(2-Aminopropyl)-5-methoxyindole Hydrochloride (6CI,7CI,8CI)
IUPAC navn 1-(5-methoxy-1H-indol-3-yl)propan-2-amin; hydrochlorid
SMIL Cl.COc1ccc2[nH]cc(CC(C)N)c2c1

Specifications

Kemisk navn eller materiale 5-Methoxy-alpha-methyltryptamine Hydrochloride
Mængde 250 mg
InChI formel InChI=1S/C12H16N2O.ClH/c1-8(13)5-9-7-14-12-4-3-10(15-2)6-11(9)12;/h3-4,6-8,14H,5,13H2,1-2H3;1H
Formel vægt 240.103
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.