Få mere at vide
Alpha-Muricholic Acid, TRC
Artikelnummer. 30138484
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30138484 10 mg
30008553 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30138484 Leverandør Toronto Research Chemicals Leverandørnr. TRCM73275010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 2393-58-0
Molekylær formel C24 H40 O5
Molekylvægt (g/mol) 408.57
Synonym Cholan-24-oic acid, 3,6,7-trihydroxy-, (3α,5β,6β,7α)-, 5β-Cholan-24-oic acid, 3α,6β,7α-trihydroxy- (8CI), 5β-Cholanic acid, 3α,6β,7α-trihydroxy- (7CI), α-Muricholic acid (6CI), (3α,5β,6β,7α)-3,6,7-Trihydroxycholan-24-oic acid, 3α,6β,7α-Trihydroxy-5β-cholan-24-oic acid, 5β-Cholanic acid-3α,6β,7α-triol, 3α,6β,7α-Trihydroxy-5β-cholanic acid
IUPAC navn (4R)-4-[(3R,5R,6S,7S,8S,9S,10R,13R,14S,17R)-3,6,7-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoic acid
SMIL C[C@H](CCC(=O)O)[C@H]1CC[C@H]2[C@@H]3[C@H](O)[C@@H](O)[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3CC[C@]12C
Tekniske data
Kemisk navn eller materiale alpha-Muricholic acid
Mængde 10 mg
InChI formel InChI=1S/C24H40O5/c1-13(4-7-19(26)27)15-5-6-16-20-17(9-11-23(15,16)2)24(3)10-8-14(25)12-18(24)21(28)22(20)29/h13-18,20-22,25,28-29H,4-12H2,1-3H3,(H,26,27)/t13-,14-,15-,16+,17+,18+,20+,21+,22+,23-,24-/m1/s1
Formel vægt 408.2876