Få mere at vide
Alpha-Methyldopa Methyl Ester, TRC
Artikelnummer. 30179731
Skift visning
Mængde:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30179731 10 mg
30180251 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30179731 Leverandør Toronto Research Chemicals Leverandørnr. TRCM30386310MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 18181-08-3
Molekylær formel C11 H15 N O4
Molekylvægt (g/mol) 225.24
Synonym Methyl (2S)-2-Amino-3-(3,4-dihydroxyphenyl)-2-methyl-propanoate, 3-Hydroxy-α-methyl-L-tyrosine Methyl Ester, Alanine, 3-(3,4-dihydroxyphenyl)-2-methyl-, methyl ester, L- (8CI), α-Methyldopa Methyl Ester
IUPAC navn methyl (2S)-2-amino-3-(3,4-dihydroxyphenyl)-2-methylpropanoate
SMIL COC(=O)[C@@](C)(N)Cc1ccc(O)c(O)c1
Tekniske data
Kemisk navn eller materiale Methyl (2S)-2-Amino-3-(3,4-dihydroxyphenyl)-2-methylpropanoate
Mængde 10 mg
InChI formel InChI=1S/C11H15NO4/c1-11(12,10(15)16-2)6-7-3-4-8(13)9(14)5-7/h3-5,13-14H,6,12H2,1-2H3/t11-/m0/s1
Formel vægt 225.1001