Få mere at vide
Alpha-Methyl-L-tryptophan, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | a-Methyl-L-tryptophan |
| CAS | 16709-25-4 |
| Molekylær formel | C12 H14 N2 O2 |
| Mængde | 10 mg |
| Synonym | L-Tryptophan, α-methyl-, Tryptophan, α-methyl-, L- (8CI), α-Methyl-L-tryptophan, (-)-α-Methyltryptophan, L-α-Methyltryptophan, α-Methyl-(S)-tryptophan, (2S)-2-Amino-3-(1H-indol-3-yl)-2-methylpropanoic acid |
| InChI formel | InChI=1S/C12H14N2O2/c1-12(13,11(15)16)6-8-7-14-10-5-3-2-4-9(8)10/h2-5,7,14H,6,13H2,1H3,(H,15,16)/t12-/m0/s1 |
| SMIL | C[C@](N)(Cc1c[nH]c2ccccc12)C(=O)O |
| IUPAC navn | (2S)-2-amino-3-(1H-indol-3-yl)-2-methylpropanoic acid |
| Molekylvægt (g/mol) | 218.25 |
| Formel vægt | 218.1055 |