Få mere at vide
alpha-Methyl-L-serine, TRC
Artikelnummer. 30005176
Skift visning
Mængde:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30005176 500 mg
30009352 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30005176 Leverandør Toronto Research Chemicals Leverandørnr. TRCM325976500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 16820-18-1
Molekylær formel C4 H9 N O3
Molekylvægt (g/mol) 119.119
Synonym (S)-(+)-2-Amino-3-hydroxy-2-methylpropionic Acid, 2-Methyl-L-serine, (S)-2-Methylserine, L-2-Methylserine, L-α-Methylserine
IUPAC navn (2S)-2-amino-3-hydroxy-2-methyl-propanoic acid
SMIL C[C@](N)(CO)C(=O)O
Tekniske data
Kemisk navn eller materiale alpha-Methyl-L-serine
InChI formel InChI=1S/C4H9NO3/c1-4(5,2-6)3(7)8/h6H,2,5H2,1H3,(H,7,8)/t4-/m0/s1
Mængde 500 mg
Formel vægt 119.0582