Få mere at vide
alpha-Lobeline Hydrochcloride, TRC
Artikelnummer. 30475460
Skift visning
Mængde:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30475460 100 mg
30472268 500 mg
30471346 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30475460 Leverandør Toronto Research Chemicals Leverandørnr. TRCL469350100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 134-63-4
Molekylær formel C22 H27 N O2 . Cl H
Molekylvægt (g/mol) 373.92
Synonym Lobeline hydrochloride, 2-[(2R,6S)-6-[(2S)-2-Hydroxy-2-phenylethyl]-1-methylpiperidin-2-yl]-1-phenylethanone hydrochloride
IUPAC navn 2-[(2R,6S)-6-[(2S)-2-hydroxy-2-phenylethyl]-1-methylpiperidin-2-yl]-1-phenylethanone;hydrochloride
SMIL Cl.CN1[C@H](C[C@H](O)c2ccccc2)CCC[C@@H]1CC(=O)c3ccccc3
Tekniske data
Kemisk navn eller materiale alpha-Lobeline hydrochloride
Mængde 100 mg
InChI formel InChI=1S/C22H27NO2.ClH/c1-23-19(15-21(24)17-9-4-2-5-10-17)13-8-14-20(23)16-22(25)18-11-6-3-7-12-18;/h2-7,9-12,19-21,24H,8,13-16H2,1H3;1H/t19-,20+,21-;/m0./s1
Formel vægt 373.1809