Få mere at vide
Alpha-Isolupanin Perchlorate, TRC
Artikelnummer. 17973895
Skift visning
Mængde:
0.5 mg
5 mg
Pakningsstørrelse:
0.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
17973895 5 mg -
17905055 0.5 mg 0.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17973895 Leverandør Toronto Research Chemicals Leverandørnr. TRCI8209005MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 14691-01-1
Molekylær formel C15H24N2O . xHClO 4
Molekylvægt (g/mol) 348.82
Synonym 11-Isolupanine Perchlorate, [7S-(7alpha,7aalpha,14alpha,14aalpha)]-Dodecahydro-7,14-Methano-2H,11H-dipyrido[1,2-a:1',2'-e][1,5]diazocin-11-one Perchlorate;
IUPAC navn (7S,7aR,14S,14aR)-11-oxotetradecahydro-2H-7,14-methanodipyrido[1,2-a:1',2'-e][1,5]diazocin-5-ium perchlorate
SMIL O=C1N2[C@@](CCC1)([H])[C@@]3([H])C[C@@]([C@]4([H])[NH+](CCCC4)C3)([H])C2.O=Cl(=O)([O-])=O
Tekniske data
Kemisk navn eller materiale (+)-alpha-Isolupanin Perchlorate
Renhedsgrad noter HPLC
Mængde 5 mg
InChI formel InChI=1S/C15H24N2O.ClHO4/c18-15-6-3-5-14-11-8-12(10-17(14)15)13-4-1-2-7-16(13)9-11;2-1(3,4)5/h11-14H,1-10H2;(H,2,3,4,5)/t11-,12-,13+,14+;/m0./s1
Formel vægt 348.1452
Procent renhed >95%