Få mere at vide
alpha-Hyodeoxycholic Acid, TRC
Artikelnummer. 30006858
Skift visning
Mængde:
5 mg
100 mg
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30006858 100 mg
30010277 5 g
30010261 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30006858 Leverandør Toronto Research Chemicals Leverandørnr. TRCH998100100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 83-49-8
Molekylær formel C24 H40 O4
Molekylvægt (g/mol) 392.57
Synonym (3alpha,5beta,6alpha)-3,6-Dihydroxycholan-24-oic Acid, 3alpha,6alpha-Dihydroxy-5beta-cholan-24-oate, 3alpha,6alpha-Dihydroxy-5beta-cholan-24-oic Acid, 6alpha-Hydroxylithocholic Acid, 7-Deoxyhyocholic Acid, Hyodeoxycholic Acid, Hyodesoxycholic Acid, Iodeoxycholic Acid, NSC 60672;
IUPAC navn (4R)-4-[(3R,5R,6S,8S,9S,10R,13R,14S,17R)-3,6-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoic acid
SMIL [H][C@@]12C[C@H](O)[C@]3([H])C[C@H](O)CC[C@]3(C)[C@@]1([H])CC[C@@]4(C)[C@@]2([H])CC[C@]4([H])[C@H](C)CCC(O)=O
Tekniske data
Kemisk navn eller materiale Hyodeoxycholic acid
Mængde 100 mg
InChI formel InChI=1S/C24H40O4/c1-14(4-7-22(27)28)17-5-6-18-16-13-21(26)20-12-15(25)8-10-24(20,3)19(16)9-11-23(17,18)2/h14-21,25-26H,4-13H2,1-3H3,(H,27,28)/t14-,15-,16+,17-,18+,19+,20+,21+,23-,24-/m1/s1
Formel vægt 392.2927