Få mere at vide
alpha-Hydroxymetoprolol (mixture of stereoisomers), CDN
Tekniske data
Tekniske data
| Kemisk navn eller materiale | a-Hydroxy Metoprolol(Mixture of Diastereomers) |
| CAS | 56392-16-6 |
| Molekylær formel | C15 H25 N O4 |
| Mængde | 0.01 g |
| Synonym | Benzenemethanol, 4-[2-hydroxy-3-[(1-methylethyl)amino]propoxy]-α-(methoxymethyl)- (9 CI, ACI), 4-[2-Hydroxy-3-[(1-methylethyl)amino]propoxy]-α-(methoxymethyl)benzenemethanol (ACI), 1-Isopropylamino-3-[4-(1-hydroxy-2-methoxyethyl)phenoxy]-2-propanol, H 119/66, α-Hydroxymetoprolol, (R,S)-α-Hydroxymetoprolol, α-Hydroxy Metoprolol, 1-[4-(1-Hydroxy-2-methoxy-ethyl)phenoxy]-3-(isopropylamino)propan-2-ol |
| InChI formel | InChI=1 S/C15H25NO4/c1-11(2)16-8-13(17)9-20-14-6-4-12(5-7-14)15(18)10-19-3/h4-7,11,13,15-18 H,8-10H2,1-3H3 |
| SMIL | COCC(O)c1ccc(OCC(O)CNC(C)C)cc1 |
| IUPAC navn | 1-[4-(1-hydroxy-2-methoxyethyl)phenoxy]-3-(propan-2-ylamino)propan-2-ol |
| Molekylvægt (g/mol) | 283.363 |
| Formel vægt | 283.178 g/mol |
| Vis mere |