Få mere at vide
alpha-Hydroxy Flurbiprofen, TRC
Artikelnummer. 30138274
Skift visning
Mængde:
250 mg
2.5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30138274 2.5 g
30006723 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30138274 Leverandør Toronto Research Chemicals Leverandørnr. TRCH9424452.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 61466-95-3
Molekylær formel C15 H13 F O3
Molekylvægt (g/mol) 260.26
Synonym (2RS)-2-(2-Fluorobiphenyl-4-yl)-2-hydroxypropanoic Acid, Flurbiprofen Imp. C (EP)
IUPAC navn 2-(3-fluoro-4-phenylphenyl)-2-hydroxypropanoic acid
SMIL CC(O)(C(=O)O)c1ccc(c(F)c1)c2ccccc2
Tekniske data
Kemisk navn eller materiale (2RS)-2-(2-Fluorobiphenyl-4-yl)-2-hydroxypropanoic Acid
Renhedsgrad noter HPLC
Mængde 2.5 g
InChI formel InChI=1S/C15H13FO3/c1-15(19,14(17)18)11-7-8-12(13(16)9-11)10-5-3-2-4-6-10/h2-9,19H,1H3,(H,17,18)
Formel vægt 260.0849
Procent renhed >95%