Få mere at vide
alpha-D-Glucopyranoside-beta-D-fructofuranosyl Octadecanoate, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | alpha-D-Glucopyranoside-beta-D-fructofuranosyl Octadecanoate |
| CAS | 37318-31-3 |
| Molekylær formel | C30 H56 O12 |
| Mængde | 10 mg |
| Synonym | α-D-Glucopyranoside β-D-fructofuranosyl octadecanoate, alpha-D-Glucopyranoside-beta-D-fructofuranosyl Octadecanoate, Sucrose, stearate (6CI), Cane sugar stearate, Cosmelike S 10, Cosmelike S 110, Crodesta F 110, Crodesta F 140, Crodesta F 160, Crodesta F 90, DK 160, DK 50, DK Ester O 10, DKF 10, DUB-SE 11S, Nitto Ester S 370, Rikemal A, Ryoto S 1170, Ryoto S 170, Ryoto Sugar Ester S 070, Ryoto Sugar Ester S 1170, Ryoto Sugar Ester S 1170F, Ryoto Sugar Ester S 1170S, Ryoto Sugar Ester S 1570, Ryoto Sugar Ester S 170, Ryoto Sugar Ester S 270, Ryoto Sugar Ester S 370F, Ryoto Sugar Ester S 395, Ryoto Sugar Ester S 570, Ryoto Sugar Ester S 70, Ryoto Sugar Ester S 970, Ryoto Sugar Ester SM 1800, S 070, S 1170, S 1570, S 170, S 270, S 395, S 970, SE 1570, SE 970, SEFA polystearate, SFE 1815, SM 90, SM 90 (sugar ester), SP 10C, SS 15, SS 7, Saccharose stearate, Sefose 1618H, Sisterna SP 50, Sucroester WE 11, Sucrose octadecanoate, Sucrose stearate, Surfhope C 1816, Surfhope J 1809, Surfhope J 1811, Surfhope J 1815, Surfhope SE COSME C 1800, Surfhope SE COSME C 1801, Surfhope SE COSME C 1802, Surfhope SE Cosme C 1811, Surfhope SE Cosme C 1815, Surfhope SE Cosme C 1816, Surfhope Sepharma J 1811F |
| InChI formel | InChI=1S/C30H56O12/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-23(33)42-30(20-32)28(38)25(35)22(41-30)19-39-29-27(37)26(36)24(34)21(18-31)40-29/h21-22,24-29,31-32,34-38H,2-20H2,1H3/t21-,22-,24-,25-,26+,27-,28+,29+,30+/m1/s1 |
| SMIL | CCCCCCCCCCCCCCCCCC(=O)O[C@]1(CO)O[C@H](CO[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O |
| IUPAC navn | [(2S,3S,4S,5R)-3,4-dihydroxy-2-(hydroxymethyl)-5-[[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxymethyl]oxolan-2-yl] octadecanoate |
| Molekylvægt (g/mol) | 608.758 |
| Formel vægt | 608.377 |