Få mere at vide
alpha-D-Galactose, TRC
Artikelnummer. 17968004
Skift visning
Mængde:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17968004 50 mg
17958964 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17968004 Leverandør Toronto Research Chemicals Leverandørnr. TRCG15526050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 3646-73-9
Molekylær formel C6 H12 O6
Molekylvægt (g/mol) 180.16
Synonym α-D-Galactopyranose, Galactopyranose, α-D- (8CI), α-D-Galactose, α-(+)-Galactose, α-D-(+)-Galactose
IUPAC navn (2S,3R,4S,5R,6R)-6-(hydroxymethyl)oxane-2,3,4,5-tetrol
SMIL OC[C@H]1O[C@H](O)[C@H](O)[C@@H](O)[C@H]1O
Tekniske data
Kemisk navn eller materiale alpha-D-Galactose
Mængde 50 mg
InChI formel InChI=1S/C6H12O6/c7-1-2-3(8)4(9)5(10)6(11)12-2/h2-11H,1H2/t2-,3+,4+,5-,6+/m1/s1
Formel vægt 180.0634