Få mere at vide
Alpha-Cholestane-d4, TRC
Artikelnummer. 30361575
Skift visning
Mængde:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30361575 5 mg
30363030 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30361575 Leverandør Toronto Research Chemicals Leverandørnr. TRCC4317025MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 205529-74-4
Molekylær formel C27 D4 H44
Molekylvægt (g/mol) 376.695
Synonym 5α-Cholestane-2,2,4,4-D4, (5α)-Cholestane-2,2,4,4-d4, α-Cholestane-d4
IUPAC navn (5R,8R,9S,10S,13R,14S,17R)-2,2,4,4-tetradeuterio-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-3,5,6,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthrene
SMIL [2H]C1([2H])CC([2H])([2H])[C@@H]2CC[C@H]3[C@@H]4CC[C@H]([C@H](C)CCCC(C)C)[C@@]4(C)CC[C@@H]3[C@@]2(C)C1
Tekniske data
Kemisk navn eller materiale alpha-Cholestane-d4
Mængde 5 mg
InChI formel InChI=1S/C27H48/c1-19(2)9-8-10-20(3)23-14-15-24-22-13-12-21-11-6-7-17-26(21,4)25(22)16-18-27(23,24)5/h19-25H,6-18H2,1-5H3/t20-,21-,22+,23-,24+,25+,26+,27-/m1/s1/i7D2,11D2
Formel vægt 376.401