missing translation for 'onlineSavingsMsg'
Få mere at vide

alfa-klorfenylacetylchlorid, TRC

Artikelnummer. 30361437
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30361437 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30361437 Leverandør Toronto Research Chemicals Leverandørnr. TRCC3765051G
 Begrænset vare
Forventet forsendelse: 10-12-2026
Læg i indkøbskurv
Læg i indkøbskurv

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy

Højrenheds organiske molekyler og analytiske standarder, strategisk leveret verden over for at muliggøre innovation og kommerciel succes.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale alpha-Chlorophenylacetyl Chloride
CAS 2912-62-1
Molekylær formel C8 H6 Cl2 O
Mængde 1 g
Synonym Chlorophenyl-acetyl Chloride, (±)-2-Chloro-2-phenylacetyl Chloride, 2-Chloro-2-phenylacetyl Chloride, 2-Phenyl-2-chloroacetyl Chloride, Chlorophenylacetyl Chloride, Phenyl(chloro)acetyl Chloride, α-Chloro-α-phenylacetyl Chloride, α-Chlorobenzeneacetyl Chloride, α-Chlorophenylacetyl Chloride
InChI formel InChI=1S/C8H6Cl2O/c9-7(8(10)11)6-4-2-1-3-5-6/h1-5,7H
SMIL ClC(C(=O)Cl)c1ccccc1
IUPAC navn 2-klor-2-fenylacetylchlorid
Molekylvægt (g/mol) 189.039
Formel vægt 187.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.