Learn More
Alpha-Cellulose (powdered), TRC
Product Code. 17997694
Change view
Click to view available options
Quantity:
50 g
100 g
500 g
Unit Size:
100g
500g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17997694 50 g 50g
30135284 500 g 500g
17999734 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17997694 Supplier Toronto Research Chemicals Supplier No. TRCC45920050G
 Restricted Item
Forventet forsendelse: 08-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 9004-34-6
Molecular Formula (C6H10O5)n
Molecular Weight (g/mol) 504.44
Synonym Cellulose, β-Amylose
IUPAC Name (2R,3R,4R,5S,6R)-6-(hydroxymethyl)-5-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)tetrahydro-2H-pyran-2,3,4-triol
SMILES O[C@@H]1[C@@H](O)[C@H](O)[C@@H](CO)O[C@H]1O[C@@H]2[C@@H](CO)O[C@@H](O[H])[C@H](O)[C@H]2O
Specifications
Quantity 50 g
InChI Formula InChI=1S/C12H22O11/c13-1-3-5(15)6(16)9(19)12(22-3)23-10-4(2-14)21-11(20)8(18)7(10)17/h3-20H,1-2H2/t3-,4-,5-,6+,7-,8-,9-,10-,11-,12+/m1/s1
Formula Weight 504.169
Chemical Name or Material Silicified Microcystalline Cellulose