Få mere at vide
Alpha-Carotene, TRC
Artikelnummer. 17947274
Skift visning
Mængde:
0.25 mg
2.5 mg
10 mg
Pakningsstørrelse:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
17947274 2.5 mg 2.5mg
17917474 0.25 mg -
17938234 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 17947274 Leverandør Toronto Research Chemicals Leverandørnr. TRCC1842452.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 432-70-2
Molekylær formel C40 H56
Molekylvægt (g/mol) 536.87
Synonym β,ε-Carotene, α-Carotene, all-trans- (8CI), (all-E)-1,3,3-Trimethyl-2-[3,7,12,16-tetramethyl-18-(2,6,6-trimethyl-2-cyclohexen-1-yl)-1,3,5,7,9,11,13,15,17-octadecanonaenyl]cyclohexene, Cyclohexene, 1,3,3-trimethyl-2-[3,7,12,16-tetramethyl-18-(2,6,6-trimethyl-2-cyclohexen-1-yl)-1,3,5,7,9,11,13,15,17-octadecanonaenyl]-, (all-E)-, Hi-Alpha, all-trans-α-Carotene, α-Carotene, rac-α-Carotene, (all-E)-1,3,3-Trimethyl-2-[3,7,12,16-tetramethyl-18-(2,6,6-trimethyl-2-cyclohexen-1-yl)-1,3,5,7,9,11,13,15,17-octadecanonaenyl]-cyclohexene, all-trans-alpha-Carotene, (all-E)-1,3,3-Trimethyl-2-[3,7,12,16-tetramethyl-18-(2,6,6-trimethyl-2-cyclohexen-1-yl)-1,3,5,7,9,11,13,15,17-octadecanonaenyl]-cyclohexene, all-trans-alpha-Carotene, alpha-Carotene, rac-alpha-Carotene
IUPAC navn 1,3,3-trimethyl-2-[(1E,3E,5E,7E,9E,11E,13E,15E,17E)-3,7,12,16-tetramethyl-18-(2,6,6-trimethylcyclohex-2-en-1-yl)octadeca-1,3,5,7,9,11,13,15,17-nonaenyl]cyclohexene
SMIL C\C(=C/C=C/C=C(\C)/C=C/C=C(\C)/C=C/C1=C(C)CCCC1(C)C)\C=C\C=C(/C)\C=C\C2C(=CCCC2(C)C)C
Tekniske data
Kemisk navn eller materiale rac-alpha-Carotene
Mængde 2.5 mg
InChI formel InChI=1S/C40H56/c1-31(19-13-21-33(3)25-27-37-35(5)23-15-29-39(37,7)8)17-11-12-18-32(2)20-14-22-34(4)26-28-38-36(6)24-16-30-40(38,9)10/h11-14,17-23,25-28,37H,15-16,24,29-30H2,1-10H3/b12-11+,19-13+,20-14+,27-25+,28-26+,31-17+,32-18+,33-21+,34-22+
Formel vægt 536.4382