Få mere at vide
Alpha-(Benzoylamino)benzeneacetamide-d10, TRC
Artikelnummer. 30340899
Skift visning
Mængde:
1 g
Artikelnummer. Mængde
30340899 1 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30340899 Leverandør Toronto Research Chemicals Leverandørnr. TRCB2079671G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale alpha-(Benzoylamino)benzeneacetamide-d10
CAS 1246814-47-0
Molekylær formel C15H4D10N2O2
Mængde 1 g
Synonym N-(alpha-Carbamoylbenzyl)benzamide-d10
InChI formel InChI=1S/C15H14N2O2/c16-14(18)13(11-7-3-1-4-8-11)17-15(19)12-9-5-2-6-10-12/h1-10,13H,(H2,16,18)(H,17,19)/i1D,2D,3D,4D,5D,6D,7D,8D,9D,10D
SMIL [2H]c1c([2H])c([2H])c(C(N=C(O)c2c([2H])c([2H])c([2H])c([2H])c2[2H])C(=N)O)c([2H])c1[2H]
IUPAC navn 2,3,4,5,6-pentadeuterio-N-[2-hydroxy-2-imino-1-(2,3,4,5,6-pentadeuteriophenyl)ethyl]benzenecarboximidic acid
Molekylvægt (g/mol) 264.35
Formel vægt 264.1683