Få mere at vide
Alpha-Artemether-d3, TRC
Artikelnummer. 30335536
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30335536 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30335536 Leverandør Toronto Research Chemicals Leverandørnr. TRCA7774121MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale alpha-Artemether-d3
CAS 93861-34-8
Molekylær formel C16H23D3O5
Mængde 1 mg
Synonym (+)-α-Artemether-d3, SM 229-d3, α-Artemether-d3, α-Dihydroartemisinin μethyl-d3 εther, (3R,5aS,6R,8aS,9R,10R,12R,12aR)-Decahydro-10-methoxy-3,6,9-trimethyl-3,12-epoxy-12H-pyrano[4,3-j]-1,2-benzodioxepin-d3
InChI formel InChI=1S/C16H26O5/c1-9-5-6-12-10(2)13(17-4)18-14-16(12)11(9)7-8-15(3,19-14)20-21-16/h9-14H,5-8H2,1-4H3/t9-,10-,11+,12+,13-,14-,15-,16-/m1/s1/i4D3
SMIL [H][C@]1(CC[C@](OO2)(C)O3)[C@H](C)CC[C@]4([H])[C@@]12[C@]3([H])O[C@@H](OC([2H])([2H])[2H])[C@@H]4C
IUPAC navn (1R,4S,5R,8S,9R,10R,12R,13R)-1,5,9-trimethyl-10-(trideuteriomethoxy)-11,14,15,16-tetraoxatetracyclo[10.3.1.04,13.08,13]hexadecane
Molekylvægt (g/mol) 301.39