Få mere at vide
alpha-Aminoisobutyric acid benzyl ester hydrochloride, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | alpha-Aminoisobutyric acid benzyl ester hydrochloride |
| CAS | 60421-20-7 |
| Molekylær formel | C11H15NO2 . HCl |
| Mængde | 250 mg |
| Synonym | Alanine, 2-methyl-, phenylmethyl ester, hydrochloride (1:1) (ACI), Alanine, 2-methyl-, phenylmethyl ester, hydrochloride (9CI), 2-Amino-2-methylpropionic acid benzyl ester hydrochloride, 2-Methyl alanine benzyl ester hydrochloride, Benzyl α-aminoisobutyrate hydrochloride, α-Aminoisobutyric acid benzyl ester hydrochloride, 2-Methylalanine phenylmethyl ester hydrochloride, 2-Amino-2-methylpropanoic acid benzyl ester hydrochloride, 2-Aminoisobutyric acid benzyl ester hydrochloride, Benzyl 2-amino-2-methylpropanoate hydrochloride, Benzyl 2-amino-2-methylpropionate hydrochloride |
| InChI formel | InChI=1S/C11H15NO2.ClH/c1-11(2,12)10(13)14-8-9-6-4-3-5-7-9;/h3-7H,8,12H2,1-2H3;1H |
| SMIL | Cl.CC(C)(N)C(=O)OCc1ccccc1 |
| IUPAC navn | benzyl 2-amino-2-methylpropanoate;hydrochloride |
| Molekylvægt (g/mol) | 229.7 |
| Formel vægt | 229.087 |