Få mere at vide
Alpha-Amino-Beta-methylaminopropionic Acid-d3 Hydrochloride, TRC
Artikelnummer. 30318637
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30318637 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30318637 Leverandør Toronto Research Chemicals Leverandørnr. TRCA6120021MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale alpha-Amino-beta-methylaminopropionic Acid-d3 Hydrochloride
CAS 1794713-10-2
Molekylær formel C4H8D3ClN2O2
Mængde 1 mg
Synonym 3-(Methyl-d3-amino)-DL-alanine Monohydrochloride, 2-Amino-3-(methyl-d3-amino)-propionic Acid Hydrochloride
InChI formel InChI=1S/C4H10N2O2.ClH/c1-6-2-3(5)4(7)8;/h3,6H,2,5H2,1H3,(H,7,8);1H/i1D3;
SMIL Cl.[2H]C([2H])([2H])NCC(N)C(=O)O
IUPAC navn 2-amino-3-(trideuteriomethylamino)propanoic acid;hydrochloride
Molekylvægt (g/mol) 157.61
Formel vægt 157.0697