Få mere at vide
alpha,alpha-Diphenyl-gamma-butyrolactone, Mikromol™
Artikelnummer. 30050782
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30050782 100 mg
30049581 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30050782 Leverandør Mikromol Leverandørnr. MM0025.08
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
alpha,alpha-Diphenyl-gamma-butyrolactone
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale alpha,alpha-Diphenyl-gamma-butyrolactone
CAS 956-89-8
Analyt- eller komponentnavne alpha,alpha-Diphenyl-gamma-butyrolactone
Forsendelsestilstand Room Temperature
Molekylær formel C16 H14 O2
Certificeringer/compliance ISO 17025
Synonym alpha,alpha-Diphenyl-gamma-butyrolactone
InChI formel InChI=1S/C16H14O2/c17-15-16(11-12-18-15,13-7-3-1-4-8-13)14-9-5-2-6-10-14/h1-10H,11-12H2
SMIL O=C1OCCC1(c2ccccc2)c3ccccc3
IUPAC navn 3,3-diphenyloxolan-2-one
Molekylvægt (g/mol) 238.28
Mængde 100 mg
Formel vægt 238.0994 g/mol