Få mere at vide
Alpha,Alpha-Diphenyl-2-pyridinemethanol Hydrochloride-d5, TRC
Artikelnummer. 30377989
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30377989 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30377989 Leverandør Toronto Research Chemicals Leverandørnr. TRCD49185725MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale alpha,alpha-Diphenyl-2-pyridinemethanol Hydrochloride-d5
CAS 1215374-19-8
Molekylær formel C18H11D5ClNO
Mængde 25 mg
Synonym A,A-Diphenyl-2-Pyridinemethanol Hydrochloride-D5
InChI formel InChI=1S/C18H15NO.ClH/c20-18(15-9-3-1-4-10-15,16-11-5-2-6-12-16)17-13-7-8-14-19-17;/h1-14,20H;1H/i1D,3D,4D,9D,10D;
SMIL Cl.[2H]c1c([2H])c([2H])c(c([2H])c1[2H])C(O)(c2ccccc2)c3ccccn3
IUPAC navn (2,3,4,5,6-pentadeuteriophenyl)-phenyl-pyridin-2-ylmethanol;hydrochloride
Molekylvægt (g/mol) 302.81
Formel vægt 302.1234