Få mere at vide
Alpha,Alpha'-Dilaurin, TRC
Artikelnummer. 30370635
Skift visning
Mængde:
250 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30370635 250 mg
30376447 1 g
30370411 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30370635 Leverandør Toronto Research Chemicals Leverandørnr. TRCD459920250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 539-93-5
Molekylær formel C27H52O5
Molekylvægt (g/mol) 456.7
Synonym Dodecanoic Acid 2-Hydroxy-1,3-propanediyl Ester, 1,3-Dilaurin, 1,3-Dilauroylglycerol, Glycerol 1,3-didodecanoate, Glycerol 1,3-dilaurate
IUPAC navn (3-dodecanoyloxy-2-hydroxypropyl) dodecanoate
SMIL CCCCCCCCCCCC(=O)OCC(O)COC(=O)CCCCCCCCCCC
Tekniske data
Kemisk navn eller materiale alpha,alpha'-Dilaurin
Mængde 250 mg
InChI formel InChI=1S/C27H52O5/c1-3-5-7-9-11-13-15-17-19-21-26(29)31-23-25(28)24-32-27(30)22-20-18-16-14-12-10-8-6-4-2/h25,28H,3-24H2,1-2H3
Formel vægt 456.3815