Få mere at vide
Alpha,Alpha,Alpha',Alpha'-Tetramethyl-5-methyl-1,3-benzenediacetonitrile-d12, TRC
Artikelnummer. 30411302
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30411302 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30411302 Leverandør Toronto Research Chemicals Leverandørnr. TRCT30361750MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale alpha,alpha,alpha',alpha'-Tetramethyl-5-methyl-1,3-benzenediacetonitrile-d12
CAS 1185098-75-2
Molekylær formel C15H6D12N2
Mængde 50 mg
Synonym α1,α1,α3,α3,5-Pentamethyl-1,3-benzenediacetonitrile-d12, 2,2'-(5-Methyl-1,3-phenylene)di(2-methylpropionitrile)-d12, 2-[3-(Cyanodimethyl methyl)-5-methylphenyl]-2-methylpropionitrile-d12, 3,5-Bis(2-cyano-2-methylethyl)toluene-d12, 3,5-Bis(2-cyanoprop-2-yl)toluene-d12
InChI formel InChI=1S/C15H18N2/c1-11-6-12(14(2,3)9-16)8-13(7-11)15(4,5)10-17/h6-8H,1-5H3/i2D3,3D3,4D3,5D3
SMIL CC1=CC(C(C#N)(C([2H])([2H])[2H])C([2H])([2H])[2H])=CC(C(C#N)(C([2H])([2H])[2H])C([2H])([2H])[2H])=C1
Molekylvægt (g/mol) 238.39
Formel vægt 238.2223