Learn More
Alpha,Alpha,Alpha’,Alpha’-Tetramethyl-5-(dibromomethyl)-1,3-benzenediacetonitrile, TRC
Product Code. 30409540
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30409540 50 mg
30412572 25 mg
30425443 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30409540 Supplier Toronto Research Chemicals Supplier No. TRCT29149550MG
 Restricted Item
Forventet forsendelse: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1027160-12-8
Molecular Formula C15 H16 Br2 N2
Molecular Weight (g/mol) 384.11
Synonym 1,3-Benzenediacetonitrile, 5-(dibromomethyl)-α1,α1,α3,α3-tetramethyl-, 5-(Dibromomethyl)-α1,α1,α3,α3-tetramethyl-1,3-benzenediacetonitrile
IUPAC Name 2-[3-(2-cyanopropan-2-yl)-5-(dibromomethyl)phenyl]-2-methylpropanenitrile
SMILES CC(C)(C#N)c1cc(cc(c1)C(C)(C)C#N)C(Br)Br
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C15H16Br2N2/c1-14(2,8-18)11-5-10(13(16)17)6-12(7-11)15(3,4)9-19/h5-7,13H,1-4H3
Formula Weight 381.968
Chemical Name or Material alpha,alpha,alpha',alpha'-Tetramethyl-5-(dibromomethyl)-1,3-benzenediacetonitrile