Få mere at vide
Alpha,Alpha-[1,1'-D2]Trehalose, TRC
Artikelnummer. 30452211
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30452211 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30452211 Leverandør Toronto Research Chemicals Leverandørnr. TRCT71870425MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale alpha,alpha-[1,1'-D2]Trehalose
CAS 1334376-67-8
Molekylær formel C12D2H20O11
Mængde 25 mg
Synonym 1-O-α-D-[1-D]Glucopyranosyl-α-D-[1-D]glucopyranoside
InChI formel InChI=1S/C12H22O11/c13-1-3-5(15)7(17)9(19)11(21-3)23-12-10(20)8(18)6(16)4(2-14)22-12/h3-20H,1-2H2/t3-,4-,5-,6-,7+,8+,9-,10-,11-,12-/m1/s1/i11D,12D
SMIL O[C@@H]1[C@@H](O)[C@H](O)[C@@H](CO)O[C@]1([2H])O[C@]2([2H])[C@H](O)[C@@H](O)[C@H](O)[C@@H](CO)O2
IUPAC navn (2R,2'R,3S,3'S,4S,4'S,5R,5'R,6R,6'R)-6,6'-oxybis(2-(hydroxymethyl)tetrahydro-2H-pyran-6-d-3,4,5-triol)
Molekylvægt (g/mol) 344.31
Formel vægt 344.13