Få mere at vide
Alpha,4-Dihydroxy-3-methoxy-Alpha-methyl-benzeneethanesulfonic Acid Potassium Salt-d5, TRC
Artikelnummer. 30376167
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30376167 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30376167 Leverandør Toronto Research Chemicals Leverandørnr. TRCD453157100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale alpha,4-Dihydroxy-3-methoxy-alpha-methyl-benzeneethanesulfonic Acid Potassium Salt-d5
CAS 1794789-29-9
Molekylær formel C10H8D5KO6S
Mængde 100 mg
Synonym alpha,4-Dihydroxy-3-methoxy-alpha-methyl-benzeneethanesulfonic Acid Monopotassium Salt-d5
InChI formel InChI=1S/C10H14O6S.K/c1-10(12,17(13,14)15)6-7-3-4-8(11)9(5-7)16-2;/h3-5,11-12H,6H2,1-2H3,(H,13,14,15);/q;+1/p-1/i1D3,6D2;
SMIL [K+].[2H]C([2H])([2H])C(O)(C([2H])([2H])c1ccc([O-])c(OC)c1)S(=O)(=O)O
IUPAC navn potassium;2-methoxy-4-(1,1,3,3,3-pentadeuterio-2-hydroxy-2-sulfopropyl)phenolate
Molekylvægt (g/mol) 305.4
Formel vægt 305.0384