missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ alpha-2B Adrenergic R/ADRA2B Recombinant Protein Antigen
Recombinant Protein
Brand: Novus Biologicals™ NBP3-17799PEP
Additional Details : Weight : 0.00970kg
Specifications
Antibody Competition | |
151 | |
100 μg | |
ADRA2B adrenergic, alpha-2B-, receptor, ADRA2L1adrenergic receptor alpha 2B, ADRA2RL1, ADRARL1, adrenergic, alpha-2B-, receptor, Alpha-2 adrenergic receptor subtype C2, alpha-2-adrenergic receptor-like 1, alpha-2B adrenergic receptor, Alpha-2B adrenoceptor, Alpha-2B adrenoreceptor, alpha-2B-adrenergic receptor, Alpha-2BAR, ALPHA2BAR, G-protein coupled receptor | |
Adaptive Immunity, GPCR, Immunology, Innate Immunity | |
alpha-2B Adrenergic R/ADRA2B |
PBS and 1M Urea, pH 7.4. | |
M.W. Theoretical: 24 kDa | |
Store at -20°C. Avoid freeze-thaw cycles. | |
ADRA2B | |
>80% by SDS-PAGE and Coomassie blue staining |