Få mere at vide
Alogliptin, TRC
Artikelnummer. 30010063
Skift visning
Mængde:
5 mg
10 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30010063 100 mg
17942975 10 mg
17953065 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30010063 Leverandør Toronto Research Chemicals Leverandørnr. TRCA575425100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 850649-61-5
Molekylær formel C18 H21 N5 O2
Synonym Benzonitrile, 2-[[6-[(3R)-3-amino-1-piperidinyl]-3,4-dihydro-3-methyl-2,4-dioxo-1(2H)-pyrimidinyl]methyl]-∣2-[[6-[(3R)-3-Amino-1-piperidinyl]-3,4-dihydro-3-methyl-2,4-dioxo-1(2H)-pyrimidinyl]methyl]benzonitrile∣(R)-2-[6-[3-Aminopiperidin-1-yl]-3-methyl-2,4-dioxo-1,2,3,4-tetrahydropyrimidin-1-ylmethyl]benzonitrile∣2-[[6-[(3R)-3-Amino-1-piperidinyl]-3,4-dihydro-3-methyl-2,4-dioxo-1(2H)-pyrimidinyl]methyl]benzonitrile∣Alogliptin
Tekniske data
Kemisk navn eller materiale Alogliptin
Mængde 100 mg
Forsendelsestilstand Room Temperature
Formel vægt 339.1695
MDL nummer MFCD09833196
Renhed >95% (HPLC)
Molekylvægt (g/mol) 339.39, 339.40
InChI nøgle ZSBOMTDTBDDKMP-OAHLLOKOSA-N
SMIL CN1C(=O)C=C(N2CCC[C@@H](N)C2)N(CC2=C(C=CC=C2)C#N)C1=O
IUPAC navn 2-({6-[(3R)-3-aminopiperidin-1-yl]-3-methyl-2,4-dioxo-1,2,3,4-tetrahydropyrimidin-1-yl}methyl)benzonitrile