missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Alobresib, MedChemExpress
Beskrivelse
Alobresib (GS-5829) is a BET bromodomain inhibitor, which represents a highly effective therapeutics agent against recurrent/chemotherapy resistant uterine serous carcinoma (USC) overexpressing c-Myc.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Alobresib |
| Fysisk form | Solid |
| CAS | 1637771-14-2 |
| Farve | Off-White |
| Mængde | 50 mg |
| Renhedsgrad noter | Research |
| Molekylær formel | C26H23N5O2 |
| Til brug med (applikation) | Cancer-programmed cell death |
| Synonym | GS-5829 |
| Opløselighedsinformation | DMSO : 83.33 mg/mL (190.47 mM; Need ultrasonic) |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?