Få mere at vide
Almotriptan-d6 Hydrochloride, TRC
Artikelnummer. 30312025
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30312025 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30312025 Leverandør Toronto Research Chemicals Leverandørnr. TRCA5752021MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Almotriptan hydrochloride N,N-dimethyl-d6
CAS 1794782-57-2
Molekylær formel C17 2H6 H19 N3 O2 S . Cl H
Mængde 1 mg
Synonym 2-[5-(Pyrrolidin-1-ylsulfonylmethyl)-1H-indol-3-yl]-N,N-bis(trideuteriomethyl)ethanamine hydrochloride, Almotriptan-D6 hydrochloride, Almotriptan-D6 hydrochloride (N,N-dimethyl D6)
InChI formel InChI=1S/C17H25N3O2S.ClH/c1-19(2)10-7-15-12-18-17-6-5-14(11-16(15)17)13-23(21,22)20-8-3-4-9-20;/h5-6,11-12,18H,3-4,7-10,13H2,1-2H3;1H/i1D3,2D3;
SMIL Cl.[2H]C([2H])([2H])N(CCc1c[nH]c2ccc(CS(=O)(=O)N3CCCC3)cc12)C([2H])([2H])[2H]
IUPAC navn 2-[5-(pyrrolidin-1-ylsulfonylmethyl)-1H-indol-3-yl]-N,N-bis(trideuteriomethyl)ethanamine;hydrochloride
Molekylvægt (g/mol) 377.96
Formel vægt 377.1811