Learn More
Allylthiourea, Pure, biological oxygen demand in water 50g, Fisher Chemical
Product Code. 10622251 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
Product Code. Quantity Unit Size
10622251 50 g 50g
1 options
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.
Product Code. 10622251 Supplier Fisher Chemical Supplier No. A/1566/47
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.
Specifications
CAS 109-57-9
Melting Point 78°C
Color Beige
pH 6.5
Packaging Plastic powder jar
Molecular Formula C4H8N2S
MDL Number 4940
Quantity 50 g
Formula Weight 116.18g/mol
Physical Form Solid
compliance-icons
Product Identifier
  • Allylthiourea
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /