missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Allylmagnesium chloride, 1M in MeTHF, Thermo Scientific Chemicals
854.54 DKK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
100 mL
500 mL
Description
Allylmagnesium chloride has been used in a study of the nucleophilic ring-opening of 2-methyleneaziridines to imines and subsequent conversion to 5,5?-disubstituted hydantoins. It is also used as a reagent for the introduction of the allyl group.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Allylmagnesium chloride has been used in a study of the nucleophilic ring-opening of 2-methyleneaziridines to imines and subsequent conversion to 5,5′-disubstituted hydantoins. It is also used as a reagent for the introduction of the allyl group.
Solubility
It reacts violently with water.
Notes
Air & moisture sensitive. Stable under recommended storage conditions.
Allylmagnesium chloride has been used in a study of the nucleophilic ring-opening of 2-methyleneaziridines to imines and subsequent conversion to 5,5′-disubstituted hydantoins. It is also used as a reagent for the introduction of the allyl group.
Solubility
It reacts violently with water.
Notes
Air & moisture sensitive. Stable under recommended storage conditions.
Specifications
Linear Formula | CH2=CHCH2MgCl |
Quantity | 100 mL |
UN Number | UN3399 |
Sensitivity | Air and moisture sensitive |
Solubility Information | It reacts violently with water. |
Formula Weight | 100.83 |
Concentration or Composition (by Analyte or Components) | 1M in MeTHF |
Chemical Name or Material | Allylmagnesium chloride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'