missing translation for 'onlineSavingsMsg'
Learn More

Allyl phenyl sulfone, 98%, Thermo Scientific Chemicals

Catalog Number p-7080741
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
16212-05-8
C9H10O2S
182.237
MFCD00014739
KYPIULIVYSQNNT-UHFFFAOYSA-N
allyl phenyl sulfone, benzene, 2-propenylsulfonyl, allylsulfonyl benzene, phenyl allyl sulfone, sulfone, allyl phenyl, allylsulphonyl benzene, allylphenyl sulfone, 3-phenylsulfonyl-1-propene, benzene, 2-propen-1-ylsulfonyl
85330
prop-2-enylsulfonylbenzene
C=CCS(=O)(=O)C1=CC=CC=C1
This item is not returnable. View return policy
16212-05-8
C9H10O2S
182.237
MFCD00014739
KYPIULIVYSQNNT-UHFFFAOYSA-N
allyl phenyl sulfone, benzene, 2-propenylsulfonyl, allylsulfonyl benzene, phenyl allyl sulfone, sulfone, allyl phenyl, allylsulphonyl benzene, allylphenyl sulfone, 3-phenylsulfonyl-1-propene, benzene, 2-propen-1-ylsulfonyl
85330
prop-2-enylsulfonylbenzene
C=CCS(=O)(=O)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 1.189
Boiling Point 111°C to 113°C (0.5 mmHg)
Flash Point >110°C (230°F)
Quantity 5 g
Beilstein 1863561
Sensitivity Light sensitive
Formula Weight 182.24
Percent Purity 98%
Chemical Name or Material Allyl phenyl sulfone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'