Få mere at vide
Allyl Phenyl Ether-d5, TRC
Artikelnummer. 30313434
Skift visning
Mængde:
250 mg
Artikelnummer. Mængde
30313434 250 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30313434 Leverandør Toronto Research Chemicals Leverandørnr. TRCA572212250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Allyl Phenyl Ether-d5
CAS 1373735-60-4
Molekylær formel C9H5D5O
Mængde 250 mg
Synonym Allyloxybenzene-d5, NSC 4746-d5, Phenyl 2-Propenyl Ether-d5, Phenyl Allyl Ether-d5, Phenylpropenyl Ether-d5, (2-Propenyloxy)benzene-d5, 1-(2-Propenyloxy)benzene-d5, 1-Phenoxy-2-propene-d5, 3-Phenoxy-1-propene-d5, 3-Phenoxypropene-d5, Ether Allyl Phenyl-d5, (2-Propenyloxy)benzene-d5
InChI formel InChI=1S/C9H10O/c1-2-8-10-9-6-4-3-5-7-9/h2-7H,1,8H2/i3D,4D,5D,6D,7D
SMIL [2H]c1c([2H])c([2H])c(OCC=C)c([2H])c1[2H]
IUPAC navn 1,2,3,4,5-pentadeuterio-6-prop-2-enoxybenzene
Molekylvægt (g/mol) 139.21
Formel vægt 139.1045