Learn More
Allyl Acetate, Sigma-Aldrich™
Product Code. 15816650
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
Product Code. Quantity Unit Size
15816650 100 mL 100mL
1 options
Product Code. 15816650 Supplier Sigma Aldrich Supplier No. 8205540100
 Restricted Item
Forventet forsendelse: 25-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 591-87-7
Grade Research
Molecular Formula CH3COOCH2CH=CH2
Molecular Weight (g/mol) 100.12
Specifications
Melting Point -96°C
Boiling Point 103°C to 104°C/1013 mbar
MDL Number MFCD00008721
Quantity 100 mL
Synonym Allyl acetate, Acetic acid allyl ester, Allyl acetate