Få mere at vide
Allocryptopine, TRC
Artikelnummer. 30137207
Skift visning
Mængde:
5 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30137207 50 mg
30132401 25 mg
30010058 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30137207 Leverandør Toronto Research Chemicals Leverandørnr. TRCA54507050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 485-91-6
Molekylær formel C21 H23 N O5
Molekylvægt (g/mol) 369.41
Synonym α-Fagarine, Thalictrimine, α-allo-Cryptopine, β-Homochelidonine
IUPAC navn 7,8-dimethoxy-11-methyl-17,19-dioxa-11-azatetracyclo[12.7.0.04,9.016,20]henicosa-1(21),4(9),5,7,14,16(20)-hexaen-2-one
SMIL COc1ccc2CC(=O)c3cc4OCOc4cc3CCN(C)Cc2c1OC
Tekniske data
Kemisk navn eller materiale alpha-Allocryptopine
Mængde 50 mg
InChI formel InChI=1S/C21H23NO5/c1-22-7-6-14-9-19-20(27-12-26-19)10-15(14)17(23)8-13-4-5-18(24-2)21(25-3)16(13)11-22/h4-5,9-10H,6-8,11-12H2,1-3H3
Formel vægt 369.1576