missing translation for 'onlineSavingsMsg'
Få mere at vide

Allo Maltol, TRC

Artikelnummer. 30310831
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30310831 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30310831 Leverandør Toronto Research Chemicals Leverandørnr. TRCA546700100MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Allo Maltol
CAS 644-46-2
Molekylær formel C6H6O3
Mængde 100 mg
Synonym 5-Hydroxy-2-methyl-4H-pyran-4-one, 2-Methyl-5-hydroxy-(4H)-pyran-4-one, 2-Methyl-5-hydroxy-1,4-pyrone, 3-Hydroxy-6-methyl-4-pyrone, 5-Hydroxy-2-methyl-gamma-pyrone, alpha-Deoxykojic Acid, NSC 166902
InChI formel InChI=1S/C6H6O3/c1-4-2-5(7)6(8)3-9-4/h2-3,8H,1H3
SMIL CC1=CC(=O)C(=CO1)O
IUPAC navn 5-hydroxy-2-methylpyran-4-one
Molekylvægt (g/mol) 126.11
Formel vægt 126.0317
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.