Få mere at vide
(all-Z)-6,9,12,15,18-Heneicosapentaenoic Acid Ethyl Ester, TRC
Artikelnummer. 30137943
Skift visning
Mængde:
1 mg
10 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30137943 1 mg
17918884 100 mg
17969624 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30137943 Leverandør Toronto Research Chemicals Leverandørnr. TRCH2456101MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 131775-86-5
Molekylær formel C23H36O2
Molekylvægt (g/mol) 344.53
Synonym (6Z,9Z,12Z,15Z,18Z)-6,9,12,15,18-Heneicosapentaenoic Acid Ethyl Ester;
IUPAC navn ethyl (6Z,9Z,12Z,15Z,18Z)-henicosa-6,9,12,15,18-pentaenoate
SMIL CC/C=C\C/C=C\C/C=C\C/C=C\C/C=C\CCCCC(OCC)=O
Tekniske data
Kemisk navn eller materiale (all-Z)-6,9,12,15,18-Heneicosapentaenoic Acid Ethyl Ester
Mængde 1 mg
InChI formel InChI=1S/C23H36O2/c1-3-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23(24)25-4-2/h5-6,8-9,11-12,14-15,17-18H,3-4,7,10,13,16,19-22H2,1-2H3/b6-5-,9-8-,12-11-,15-14-,18-17-
Formel vægt 344.2715