Få mere at vide
all-trans-Retinoic Acid Methyl Ester, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | all-trans Retinoic Acid Methyl Ester |
| CAS | 339-16-2 |
| Molekylær formel | C21 H30 O2 |
| Mængde | 5 mg |
| Synonym | Retinoic acid','methyl ester, Retinoic acid','methyl ester','all-trans- (8CI), Methyl all-trans-retinoate, Methyl retinoate, Ro 4-3781, Vitamin A acid methyl ester, all-trans-Methyl retinoate, all-trans-Retinoic acid methyl ester, all-trans-Vitamin A acid methyl ester, β-Retinoic acid methyl ester |
| InChI formel | InChI=1S/C21H30O2/c1-16(9-7-10-17(2)15-20(22)23-6)12-13-19-18(3)11-8-14-21(19,4)5/h7,9-10,12-13,15H,8,11,14H2,1-6H3/b10-7+,13-12+,16-9+,17-15+ |
| SMIL | COC(=O)\C=C(/C)\C=C\C=C(/C)\C=C\C1=C(C)CCCC1(C)C |
| IUPAC navn | methyl (2E,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohexen-1-yl)nona-2,4,6,8-tetraenoate |
| Molekylvægt (g/mol) | 314.46 |
| Formel vægt | 314.2246 |