Få mere at vide
Eurofins DiscoverX ALK1 Protein, Active
Beskrivelse
ALK1 is a type I cell-surface receptor for the TGF-beta superfamily of ligands. It shares with other type I receptors a high degree of similarity in serine-threonine kinase subdomains, a glycine- and serine-rich region (called the GS domain) preceding the kinase domain, and a short C-terminal tail. The encoded protein, sometimes termed ALK1, shares similar domain structures with other closely related ALK or activin receptor-like kinase proteins that form a subfamily of receptor serine/threonine kinases. Mutations in this gene are associated with hemorrhagic telangiectasia type 2, also known as Rendu-Osler-Weber syndrome 2.
GenBank L17075.1
Tekniske data
Tekniske data
| Formulering | mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 300 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution. |
| Molekylvægt (g/mol) | 45 kDa |
| Mængde | 250 μg |
| Arter | Human |
| Rekombinant | Recombinant |
| Protein tag | N-terminal 6His |
| Udtrykssystem | Recombinant enzyme expressed in Sf21 insect cells |
| Protein form | Truncated |
| Renhed eller kvalitetsklasse | ≥70% |
| Protein | ALK1, active |
Fisher Scientific Encompass-programmet tilbyder varer, som ikke er en del af vores distributionsportefølje. Disse produkter har typisk ikke billeder eller detaljerede beskrivelser. Vi er dog forpligtet til at forbedre din shoppingoplevelse. Brug formularen nedenfor til at give feedback relateret til indholdet på dette produkt.