missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Aliskiren Hemifumarate, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Aliskiren Hemifumarate |
| CAS | 173334-58-2 |
| Molekylær formel | 2 C30 H53 N3 O6 . C4 H4 O4 |
| Mængde | 2 mg |
| Synonym | (alphaS,gammaS,deltaS,zetaS)-delta-Amino-N-(3-amino-2,2-dimethyl-3-oxopropyl)-gamma-hydroxy-4-methoxy-3-(3-methoxypropoxy)-alpha,zeta-bis(1-methylethyl)benzeneoctanamide (2E)-2-butenedioate (2:1), Benzeneoctanamide, delta-amino-N-(3-amino-2,2-dimethyl-3-oxopropyl)-γ-hydroxy-4-methoxy-3-(3-methoxypropoxy)-alpha,zeta-bis(1-methylethyl)-, (alphaS,gammaS,deltaS,zetaS)-, (2E)-2-butenedioate (2:1) (salt) (9CI), Benzeneoctanamide, delta-amino-N-(3-amino-2,2-dimethyl-3-oxopropyl)-gamma-hydroxy-4-methoxy-3-(3-methoxypropoxy)-alpha,zeta-bis(1-methylethyl)-, [alphaS-(alphaR*,gammaR*,deltaR*,zetaR*)]-, (E)-2-butenedioate (2:1) (salt), Aliskiren fumarate, Aliskiren hemifumarate |
| InChI formel | InChI=1S/2C30H53N3O6.C4H4O4/c2*1-19(2)22(14-21-10-11-26(38-8)27(15-21)39-13-9-12-37-7)16-24(31)25(34)17-23(20(3)4)28(35)33-18-30(5,6)29(32)36;5-3(6)1-2-4(7)8/h2*10-11,15,19-20,22-25,34H,9,12-14,16-18,31H2,1-8H3,(H2,32,36)(H,33,35);1-2H,(H,5,6)(H,7,8)/b;;2-1+/t2*22-,23-,24-,25-;/m00./s1 |
| SMIL | COCCCOc1cc(C[C@@H](C[C@H](N)[C@@H](O)C[C@@H](C(C)C)C(=O)NCC(C)(C)C(=O)N)C(C)C)ccc1OC.COCCCOc2cc(C[C@@H](C[C@H](N)[C@@H](O)C[C@@H](C(C)C)C(=O)NCC(C)(C)C(=O)N)C(C)C)ccc2OC.OC(=O)\C=C\C(=O)O |
| IUPAC navn | (2S,4S,5S,7S)-5-amino-N-(3-amino-2,2-dimethyl-3-oxopropyl)-4-hydroxy-7-[[4-methoxy-3-(3-methoxypropoxy)phenyl]methyl]-8-methyl-2-propan-2-ylnonanamide;(E)-but-2-enedioic acid |
| Molekylvægt (g/mol) | 1219.59 |
| Formel vægt | 1218.7978 |
Produkttitel
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?