Få mere at vide
Aliskiren-d6 Hydrochloride, TRC
Artikelnummer. 30324712
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30324712 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30324712 Leverandør Toronto Research Chemicals Leverandørnr. TRCA5360021MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Aliskiren-d6 hydrochloride
CAS 1246815-96-2
Molekylær formel C30 D6 H47 N3 O6 . H Cl
Mængde 1 mg
Synonym (αS,γS,δS,ζS)-δ-Amino-N-(3-amino-2,2-dimethyl-3-oxopropyl)-γ-hydroxy-4-methoxy-3-(3-methoxypropoxy)-α,z-bis(1-methylethyl)benzeneoctanamide-d6 Hydrochloride, CGP 60536-d6, CGP60536B-d6, Rasilez-d6 Hydrochloride, SPP 100-d6, Tekturna Hydrochloride-d6
InChI formel InChI=1S/C30H53N3O6.ClH/c1-19(2)22(14-21-10-11-26(38-8)27(15-21)39-13-9-12-37-7)16-24(31)25(34)17-23(20(3)4)28(35)33-18-30(5,6)29(32)36;/h10-11,15,19-20,22-25,34H,9,12-14,16-18,31H2,1-8H3,(H2,32,36)(H,33,35);1H/t22-,23-,24-,25-;/m0./s1/i5D3,6D3;
SMIL Cl.[2H]C([2H])([2H])C(CNC(=O)[C@@H](C[C@H](O)[C@@H](N)C[C@H](Cc1ccc(OC)c(OCCCOC)c1)C(C)C)C(C)C)(C(=O)N)C([2H])([2H])[2H]
IUPAC navn (2S,4S,5S,7S)-5-amino-N-[2-carbamoyl-3,3,3-trideuterio-2-(trideuteriomethyl)propyl]-4-hydroxy-2-isopropyl-7-[[4-methoxy-3-(3-methoxypropoxy)phenyl]methyl]-8-methyl-nonanamide;hydrochloride
Molekylvægt (g/mol) 594.26
Formel vægt 583.4078