Få mere at vide
Algestone Acetophenide, TRC
Artikelnummer. 17960655
Skift visning
Mængde:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17960655 1 g
17913075 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17960655 Leverandør Toronto Research Chemicals Leverandørnr. TRCA5320401G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 24356-94-3
Molekylær formel C29 H36 O4
Synonym Pregn-4-ene-3,20-dione, 16,17-[[(1R)-1-phenylethylidene]bis(oxy)]-, (16α)-∣Pregn-4-ene-3,20-dione, 16,17-[(1-phenylethylidene)bis(oxy)]-, [16α(R)]-∣Pregn-4-ene-3,20-dione, 16α,17-dihydroxy-, cyclic acetal with acetophenone, (R)- (8CI)∣2H-Naphth[1',2':6,7]indeno[1,2-d][1,3]dioxole, pregn-4-ene-3,20-dione deriv.∣Algestone acetophenide∣Alphasone acetophenide∣Deladroxone∣Dihydroxyprogesterone acetophenide∣Droxone∣Neolutin Depositum∣P-DHP∣ZK 9349∣2H-Naphth[1',2':6,7]indeno[1,2-d][1,3]dioxole, pregn-4-ene-3,20-dione deriv.∣Algestone acetophenide∣Alphasone acetophenide∣Deladroxone∣Dihydroxyprogesterone acetophenide∣Droxone∣Neolutin Depositum∣P-DHP∣ZK 9349∣Pregn-4-ene-3,20-dione, 16,17-[[(1R)-1-phenylethylidene]bis(oxy)]-, (16alpha)-∣Pregn-4-ene-3,20-dione, 16,17-[(1-phenylethylidene)bis(oxy)]-, [16alpha(R)]-∣Pregn-4-ene-3,20-dione, 16alpha,17-dihydroxy-, cyclic acetal with acetophenone, (R)- (8CI)
Tekniske data
Kemisk navn eller materiale Algestone Acetophenide
Mængde 1 g
Forsendelsestilstand Room Temperature
Formel vægt 448.2614
Molekylvægt (g/mol) 448.59